1-Propylpyrazol-4-ol 98%
Supplier: Apollo Scientific
OR960050-1G OR960050-5G
APOSOR960050-1GEA 270 EUR
APOSOR960050-1G APOSOR960050-5G
1-Propylpyrazol-4-ol 98%
1-Propylpyrazol-4-ol
Formula: C₆H₁₀N₂O
MW: 126.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22192551
CAS Number: 78242-21-4