3-(Propane-2-sulphonyl)aniline 98%
Supplier: Apollo Scientific
OR960081-1G OR960081-5G
APOSOR960081-1GEA 219 EUR
APOSOR960081-1G APOSOR960081-5G
3-(Propane-2-sulphonyl)aniline 98%
3-(Propane-2-sulphonyl)aniline
Formula: C₉H₁₃NO₂S
Storage Temperature: Ambient
MDL Number: MFCD09909519
CAS Number: 170856-37-8