2-Thiazol-2-yl-piperidine 95%
Supplier: Apollo Scientific
OR961629-100MG OR961629-1G OR961629-250MG
APOSOR961629-100MGEA 145 EUR
APOSOR961629-100MG APOSOR961629-1G APOSOR961629-250MG
2-Thiazol-2-yl-piperidine 95%
2-Thiazol-2-yl-piperidine
Formula: C₈H₁₂N₂S
Storage Temperature: Ambient
MDL Number: MFCD04966897
CAS Number: 526183-08-4