2-(4-1,2,3-Thiadiazolyl)pyridine 95%
Supplier: Apollo Scientific
OR962424-1G OR962424-250MG
APOSOR962424-1GEA 270 EUR
APOSOR962424-1G APOSOR962424-250MG
2-(4-1,2,3-Thiadiazolyl)pyridine 95%
2-(4-1,2,3-Thiadiazolyl)pyridine
Formula: C₇H₅N₃S
Storage Temperature: Ambient
MDL Number: MFCD01109388
CAS Number: 176037-42-6