2-(2-Hydroxyethoxy)-benzonitrile 98%
Supplier: Apollo Scientific
OR962753-1G OR962753-5G
APOSOR962753-1GEA 212 EUR
APOSOR962753-1G APOSOR962753-5G
2-(2-Hydroxyethoxy)-benzonitrile 98%
2-(2-Hydroxyethoxy)-benzonitrile
Formula: C₉H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD09743449
CAS Number: 313655-45-7