2,4-Dimethylpyrrole-3-carboxylic acid 98%
Supplier: Apollo Scientific
OR962756-10G OR962756-1G OR962756-5G
APOSOR962756-10GEA 592 EUR
APOSOR962756-10G APOSOR962756-1G APOSOR962756-5G
2,4-Dimethylpyrrole-3-carboxylic acid 98%
2,4-Dimethylpyrrole-3-carboxylic acid
Formula: C₇H₉NO₂
MW: 139.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750426
CAS Number: 17106-13-7