(3-Methoxy-phenyl)-oxo-acetonitrile 95%
Supplier: Apollo Scientific
OR963258-1G OR963258-5G
APOSOR963258-1GEA 226 EUR
APOSOR963258-1G APOSOR963258-5G
(3-Methoxy-phenyl)-oxo-acetonitrile 95%
(3-Methoxy-phenyl)-oxo-acetonitrile
Formula: C₉H₇NO₂
MW: 161.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658461
CAS Number: 23194-66-3