2,2-Dimethyl-chroman-4-one 97%
Supplier: Apollo Scientific
Synonyms: 2,2-Dimethylchroman-4-one
OR963494-1G OR963494-250MG
APOSOR963494-1GEA 538 EUR
APOSOR963494-1G APOSOR963494-250MG
2,2-Dimethyl-chroman-4-one 97%
2,2-Dimethyl-chroman-4-one
Formula: C₁₁H₁₂O₂
MW: 176.212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369943
CAS Number: 3780-33-4