4-Amino-6-piperidinopyrimidine 95%

Supplier: Apollo Scientific

Synonyms: 6-(Piperidin-1-yl)pyrimidin-4-amine

OR964193-1G OR964193-250MG
APOSOR964193-1GEA 136 EUR
APOSOR964193-1G APOSOR964193-250MG
4-Amino-6-piperidinopyrimidine 95%
4-Amino-6-piperidinopyrimidine
Formula: C₉H₁₄N₄
Storage Temperature: Ambient
MDL Number: MFCD09999227
CAS Number: 69206-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR