1-(2,5-Dimethoxyphenyl)ethanol 95%
Supplier: Apollo Scientific
OR965059-1G OR965059-5G
APOSOR965059-1GEA 36.4 EUR
APOSOR965059-1G APOSOR965059-5G
1-(2,5-Dimethoxyphenyl)ethanol 95%
1-(2,5-Dimethoxyphenyl)ethanol
Formula: C₁₀H₁₄O₃
MW: 182.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08442838
CAS Number: 41038-40-8