Tris{tris[3,5-bis(trifluoromethyl)phenyl]phosphine}palladium
Supplier: Apollo Scientific
Warning
Synonyms: Superstable Pd(0) catalyst
PC10715-5G PC10715-1G
APOSPC10715-5GEA 319 EUR
APOSPC10715-5G APOSPC10715-1G
Tris{tris[3,5-bis(trifluoromethyl)phenyl]phosphine}palladium
Tris{tris[3,5-bis(trifluoromethyl)phenyl]phosphine}palladium
Formula: Pd·3C₂₄H₉F₁₈P
MW: 2117.24 g/mol
Melting Pt: 169.5 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD25372942
CAS Number: 1130784-80-3