5-(2-Fluorophenyl)-2-furoic acid 95%
Supplier: Apollo Scientific
Warning
Synonyms: 5-(2-Fluorophenyl)furan-2-carboxylic acid, 2-Carboxy-5-(2-fluorophenyl)furan
PC11151-250MG PC11151-500MG
APOSPC11151-250MGEA 301 EUR
APOSPC11151-250MG APOSPC11151-500MG
5-(2-Fluorophenyl)-2-furoic acid 95%
5-(2-Fluorophenyl)-2-furoic acid
Formula: C₁₁H₇FO₃
MW: 206.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02221394
CAS Number: 353761-02-1