4-Chloro-6-(4-fluorophenyl)thieno[3,2-d]pyrimidine
Supplier: Apollo Scientific
Warning
PC11265-1G PC11265-250MG
APOSPC11265-1GEA 326 EUR
APOSPC11265-1G APOSPC11265-250MG
4-Chloro-6-(4-fluorophenyl)thieno[3,2-d]pyrimidine
4-Chloro-6-(4-fluorophenyl)thieno[3,2-d]pyrimidine
Formula: C₁₂H₆ClFN₂S
MW: 264.706 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01312344
CAS Number: 681260-56-0