Hexakis(2,2-difluoroethoxy)phosphazene 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2,4,4,6,6-Hexakis(2,2-difluoroethoxy)-1,3,5-triazatriphosphinine
PC1165-1G PC1165-250MG
APOSPC1165-1GEA 654 EUR
APOSPC1165-1G APOSPC1165-250MG
Hexakis(2,2-difluoroethoxy)phosphazene 95%
Hexakis(2,2-difluoroethoxy)phosphazene
Formula: C₁₂H₁₈F₁₂N₃O₆P₃
MW: 621.19 g/mol
Melting Pt: 41…42 °C
Storage Temperature: Ambient
MDL Number: MFCD07784333
CAS Number: 186817-57-2