1,4-Bis(heptafluoroisopropyl)benzene 97%
Supplier: Apollo Scientific
Warning
PC1227K-5G PC1227K-1G
APOSPC1227K-5GEA 661 EUR
APOSPC1227K-5G APOSPC1227K-1G
1,4-Bis(heptafluoroisopropyl)benzene 97%
1,4-Bis(heptafluoroisopropyl)benzene
Formula: C₁₂H₄F₁₄
MW: 414.138 g/mol
Melting Pt: 61…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00041486
CAS Number: 51114-12-6