7-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
Supplier: Apollo Scientific
PC1931G-1G PC1931G-5G
APOSPC1931G-1GEA 123 EUR
APOSPC1931G-1G APOSPC1931G-5G
7-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
7-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃NO
Melting Pt: >280 °C
Storage Temperature: Ambient
MDL Number: MFCD00126367
CAS Number: 57124-20-6