Ethyl 1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate 97%
Supplier: Apollo Scientific
PC200107-1G PC200107-250MG
APOSPC200107-1GEA 406 EUR
APOSPC200107-1G APOSPC200107-250MG
Ethyl 1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate 97%
Ethyl 1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
Formula: C₈H₉F₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18262320
CAS Number: 852228-09-2