2-(4-Fluorophenyl)acetohydrazide 97%
Supplier: Apollo Scientific
PC200536-5G PC200536-1G
APOSPC200536-5GEA 43.5 EUR
APOSPC200536-5G APOSPC200536-1G
2-(4-Fluorophenyl)acetohydrazide 97%
2-(4-Fluorophenyl)acetohydrazide
Formula: C₈H₉FN₂O
MW: 168.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06655053
CAS Number: 34547-28-9