3-(4-(Bromomethyl)-3-fluorophenyl)-1,2,4-oxadiazole
Supplier: Apollo Scientific
Danger
PC201027-100MG PC201027-250MG
APOSPC201027-100MGEA 88.6 EUR
APOSPC201027-100MG APOSPC201027-250MG
3-(4-(Bromomethyl)-3-fluorophenyl)-1,2,4-oxadiazole
3-(4-(Bromomethyl)-3-fluorophenyl)-1,2,4-oxadiazole
Formula: C₉H₆BrFN₂O
Melting Pt: 87.3 °C
Storage Temperature: Ambient
MDL Number: MFCD13181201
CAS Number: 1146699-64-0
UN: 3261
ADR: 8,III