3-Fluoro-2-methoxyisonicotinonitrile 98%
Supplier: Apollo Scientific
PC201058-100MG
PC201058-1G
PC201058-250MG
APOSPC201058-100MGEA
203
EUR
APOSPC201058-100MG
APOSPC201058-1G
APOSPC201058-250MG
3-Fluoro-2-methoxyisonicotinonitrile 98%
3-Fluoro-2-methoxyisonicotinonitrile
Formula:
C₇H₅FN₂O MW: 152.126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27997090 CAS Number: 1210041-69-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...