1-(4-(Trifluoromethyl)phenyl)propan-2-one 95+%
Supplier: Apollo Scientific
PC201290-1G PC201290-5G
APOSPC201290-1GEA 104 EUR
APOSPC201290-1G APOSPC201290-5G
1-(4-(Trifluoromethyl)phenyl)propan-2-one 95+%
1-(4-(Trifluoromethyl)phenyl)propan-2-one
Formula: C₁₀H₉F₃O
MW: 202.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045104
CAS Number: 713-45-1