2-Chloro-5-[8-chloro-6-(trifluoromethyl)imidazo[1,5-a]pyridin-3-yl]pyridine
Supplier: Apollo Scientific
PC210069-1G PC210069-500MG
APOSPC210069-1GEA 562 EUR
APOSPC210069-1G APOSPC210069-500MG
2-Chloro-5-[8-chloro-6-(trifluoromethyl)imidazo[1,5-a]pyridin-3-yl]pyridine
2-Chloro-5-[8-chloro-6-(trifluoromethyl)imidazo[1,5-a]pyridin-3-yl]pyridine
Formula: C₁₃H₆Cl₂F₃N₃
MW: 332.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28127035
CAS Number: 1630096-66-0