2-Fluoro-5-nitrobenzoyl chloride
Supplier: Apollo Scientific
Danger
PC2622-1G PC2622-5G
APOSPC2622-1GEA 94.5 EUR
APOSPC2622-1G APOSPC2622-5G
2-Fluoro-5-nitrobenzoyl chloride
2-Fluoro-5-nitrobenzoyl chloride
Formula: C₇H₃ClFNO₃
Boiling Pt: 128…130 °C (2.5 mm Hg)
Melting Pt: 60…61 °C
Storage Temperature: Ambient
MDL Number: MFCD08458077
CAS Number: 709-46-6
UN: 3261
ADR: 8,III