5-Chloro-2-nitro-4-(trifluoromethoxy)aniline 98%
Supplier: Apollo Scientific
Warning
PC2821-10G PC2821-1G PC2821-5G
APOSPC2821-10GEA 282 EUR
APOSPC2821-10G APOSPC2821-1G APOSPC2821-5G
5-Chloro-2-nitro-4-(trifluoromethoxy)aniline 98%
5-Chloro-2-nitro-4-(trifluoromethoxy)aniline
Formula: C₇H₄ClF₃N₂O₃
MW: 256.566 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972735
CAS Number: 914637-23-3