4-Chloro-N-((dimethylamino)methylene)-2-fluorobenzamide
Supplier: Apollo Scientific
Warning
PC28282-1G PC28282-250MG
APOSPC28282-1GEA 104 EUR
APOSPC28282-1G APOSPC28282-250MG
4-Chloro-N-((dimethylamino)methylene)-2-fluorobenzamide
4-Chloro-N-((dimethylamino)methylene)-2-fluorobenzamide
Formula: C₁₀H₁₀ClFN₂O
MW: 228.651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31812109
CAS Number: 2379917-17-4