2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)-2,2-difluoroacetohydrazide
Supplier: Apollo Scientific
Warning
PC28290-1G PC28290-250MG PC28290-5G
APOSPC28290-1GEA 731 EUR
APOSPC28290-1G APOSPC28290-250MG APOSPC28290-5G
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)-2,2-difluoroacetohydrazide
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)-2,2-difluoroacetohydrazide
Formula: C₈H₅ClF₅N₃O
MW: 289.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29042511
CAS Number: 1823183-73-8