3-[(2-Fluorophenyl)amino]propanoic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Amino-N-(2-fluorophenyl)propionic acid
PC2927-1G PC2927-5G PC2927-250MG
APOSPC2927-1GEA 89.9 EUR
APOSPC2927-1G APOSPC2927-5G APOSPC2927-250MG
3-[(2-Fluorophenyl)amino]propanoic acid 97%
3-[(2-Fluorophenyl)amino]propanoic acid
Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
Melting Pt: 100…101 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07784302
CAS Number: 38470-19-8