8-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
PC300502-1G PC300502-250MG
APOSPC300502-1GEA 173 EUR
APOSPC300502-1G APOSPC300502-250MG
8-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine
8-Bromo-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD11847631
CAS Number: 1277178-00-3