2,6-Difluoro-4-(morpholin-4-yl)benzonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 4-(4-Cyano-3,5-difluorophenyl)morpholine
PC300690-1G PC300690-250MG
APOSPC300690-1GEA 546 EUR
APOSPC300690-1G APOSPC300690-250MG
2,6-Difluoro-4-(morpholin-4-yl)benzonitrile
2,6-Difluoro-4-(morpholin-4-yl)benzonitrile
Formula: C₁₁H₁₀F₂N₂O
MW: 224.207 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD19442761
CAS Number: 840481-30-3