Methyl 3,4-difluoro-5-(trifluoromethyl)benzoate
Supplier: Apollo Scientific
Warning
PC300747-1G PC300747-250MG
APOSPC300747-1GEA 655 EUR
APOSPC300747-1G APOSPC300747-250MG
Methyl 3,4-difluoro-5-(trifluoromethyl)benzoate
Methyl 3,4-difluoro-5-(trifluoromethyl)benzoate
Formula: C₉H₅F₅O₂
MW: 240.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042814
CAS Number: 1427460-23-8