Methyl 2,3,5-trifluoro-4-morpholin-4-ylbenzoate
Supplier: Apollo Scientific
PC300761-1G PC300761-250MG
APOSPC300761-1GEA 232 EUR
APOSPC300761-1G APOSPC300761-250MG
Methyl 2,3,5-trifluoro-4-morpholin-4-ylbenzoate
Methyl 2,3,5-trifluoro-4-morpholin-4-ylbenzoate
Formula: C₁₂H₁₂F₃NO₃
MW: 275.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27995633
CAS Number: 1858250-71-1