3-[3-(Trifluoromethoxy)phenyl]-1H-pyrazole
Supplier: Apollo Scientific
PC300781-1G PC300781-250MG PC300781-5G
APOSPC300781-1GEA 232 EUR
APOSPC300781-1G APOSPC300781-250MG APOSPC300781-5G
3-[3-(Trifluoromethoxy)phenyl]-1H-pyrazole
3-[3-(Trifluoromethoxy)phenyl]-1H-pyrazole
Formula: C₁₀H₇F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD26939177
CAS Number: 149739-45-7