2,3,5-Trifluoro-4-pyrrolidin-1-ylbenzoic acid
Supplier: Apollo Scientific
PC300783-1G PC300783-250MG
APOSPC300783-1GEA 348 EUR
APOSPC300783-1G APOSPC300783-250MG
2,3,5-Trifluoro-4-pyrrolidin-1-ylbenzoic acid
2,3,5-Trifluoro-4-pyrrolidin-1-ylbenzoic acid
Formula: C₁₁H₁₀F₃NO₂
MW: 245.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103345
CAS Number: 150660-42-7