Ethyl (4-fluorophenyl)(hydroxy)acetate
Supplier: Apollo Scientific
PC300965-1G PC300965-50MG PC300965-250MG PC300965-500MG PC300965-2.5G
APOSPC300965-1GEA 397 EUR
APOSPC300965-1G APOSPC300965-50MG APOSPC300965-250MG APOSPC300965-500MG APOSPC300965-2.5G
Ethyl (4-fluorophenyl)(hydroxy)acetate
Ethyl (4-fluorophenyl)(hydroxy)acetate
Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16661866
CAS Number: 7550-03-0