2,3-Dichloro-6-fluorobenzamide 97%
Supplier: Apollo Scientific
PC302222-10G PC302222-5G PC302222-1G
APOSPC302222-10GEA 328 EUR
APOSPC302222-10G APOSPC302222-5G APOSPC302222-1G
2,3-Dichloro-6-fluorobenzamide 97%
2,3-Dichloro-6-fluorobenzamide
Formula: C₇H₄Cl₂FNO
MW: 208.017 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115877
CAS Number: 886497-63-8