3-Fluoro-4-(trifluoromethoxy)benzamide 98%
Supplier: Apollo Scientific
PC302550-1G PC302550-5G
APOSPC302550-1GEA 116 EUR
APOSPC302550-1G APOSPC302550-5G
3-Fluoro-4-(trifluoromethoxy)benzamide 98%
3-Fluoro-4-(trifluoromethoxy)benzamide
Formula: C₈H₅F₄NO₂
MW: 223.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660208
CAS Number: 886499-18-9