3,5-Difluoro-D-phenylalanine, N-BOC protected
Supplier: Apollo Scientific
Synonyms: Boc-D-3,5-Difluorophenylalanine
PC3026-1G PC3026-25G PC3026-5G
APOSPC3026-1GEA 35.1 EUR
APOSPC3026-1G APOSPC3026-25G APOSPC3026-5G
3,5-Difluoro-D-phenylalanine, N-BOC protected
3,5-Difluoro-D-phenylalanine, N-BOC protected
Formula: C₁₄H₁₇F₂NO₄
MW: 301.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00797556
CAS Number: 205445-53-0