4-Chloro-3-fluoro-5-(trifluoromethyl)phenylacetic acid 98%
Supplier: Apollo Scientific
PC302870-1G PC302870-5G
APOSPC302870-1GEA 249 EUR
APOSPC302870-1G APOSPC302870-5G
4-Chloro-3-fluoro-5-(trifluoromethyl)phenylacetic acid 98%
4-Chloro-3-fluoro-5-(trifluoromethyl)phenylacetic acid
Formula: C₉H₅ClF₄O₂
MW: 256.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23795336
CAS Number: 1431329-73-5