2,3-Dichloro-6-(trifluoromethyl)benzoyl chloride
Supplier: Apollo Scientific
PC302905-10G PC302905-5G
APOSPC302905-10GEA 405 EUR
APOSPC302905-10G APOSPC302905-5G
2,3-Dichloro-6-(trifluoromethyl)benzoyl chloride
2,3-Dichloro-6-(trifluoromethyl)benzoyl chloride
Formula: C₈H₂Cl₃F₃O
MW: 277.455 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660271
CAS Number: 886501-87-7