2-Fluoro-5-methyl-3-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC303145-1G PC303145-5G
APOSPC303145-1GEA 339 EUR
APOSPC303145-1G APOSPC303145-5G
2-Fluoro-5-methyl-3-(trifluoromethyl)benzaldehyde 98%
2-Fluoro-5-methyl-3-(trifluoromethyl)benzaldehyde
Formula: C₉H₆F₄O
MW: 206.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054185
CAS Number: 1706458-33-4
UN: 2811
ADR: 6.1 II