2-Fluoro-4-methyl-3-(trifluoromethyl)benzoyl chloride 98%
Supplier: Apollo Scientific
PC303168-1G PC303168-250MG
APOSPC303168-1GEA 620 EUR
APOSPC303168-1G APOSPC303168-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)benzoyl chloride 98%
2-Fluoro-4-methyl-3-(trifluoromethyl)benzoyl chloride
Formula: C₉H₅ClF₄O
MW: 240.582 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103362
CAS Number: 1824271-04-6