3-Methoxy-5-(trifluoromethyl)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
PC303195-1G PC303195-250MG
APOSPC303195-1GEA 696 EUR
APOSPC303195-1G APOSPC303195-250MG
3-Methoxy-5-(trifluoromethyl)benzenesulphonyl chloride 97%
3-Methoxy-5-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₈H₆ClF₃O₃S
MW: 274.645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072526
CAS Number: 1146355-33-0