3,4-Dichloro-5-(trifluoromethoxy)benzyl bromide 97%
Supplier: Apollo Scientific
PC303196-1G PC303196-250MG
APOSPC303196-1GEA 765 EUR
APOSPC303196-1G APOSPC303196-250MG
3,4-Dichloro-5-(trifluoromethoxy)benzyl bromide 97%
3,4-Dichloro-5-(trifluoromethoxy)benzyl bromide
Formula: C₈H₄BrCl₂F₃O
MW: 323.922 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054207
CAS Number: 1706435-11-1
UN: 3265
ADR: 8 III