2,6-Difluoro-4-methoxybenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
PC303257-1G PC303257-100MG PC303257-250MG
APOSPC303257-1GEA 509 EUR
APOSPC303257-1G APOSPC303257-100MG APOSPC303257-250MG
2,6-Difluoro-4-methoxybenzenesulphonyl chloride
2,6-Difluoro-4-methoxybenzenesulphonyl chloride
Formula: C₇H₅ClF₂O₃S
MW: 242.628 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12813878
CAS Number: 1178498-86-6
UN: 1759
ADR: 8 III