2'-Fluoro-3'-methyl-5'-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
PC303384-1G PC303384-250MG
APOSPC303384-1GEA 650 EUR
APOSPC303384-1G APOSPC303384-250MG
2'-Fluoro-3'-methyl-5'-(trifluoromethyl)acetophenone 97%
2'-Fluoro-3'-methyl-5'-(trifluoromethyl)acetophenone
Formula: C₁₀H₈F₄O
MW: 220.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31645673
CAS Number: 2055275-11-9