2-Fluoro-4-methyl-5-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
PC303388-1G PC303388-5G
APOSPC303388-1GEA 430 EUR
APOSPC303388-1G APOSPC303388-5G
2-Fluoro-4-methyl-5-(trifluoromethyl)benzaldehyde 98%
2-Fluoro-4-methyl-5-(trifluoromethyl)benzaldehyde
Formula: C₉H₆F₄O
MW: 206.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27927223
CAS Number: 669080-88-0