4'-Chloro-2'-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
Synonyms: 1-(4-Chloro-2-(trifluoromethyl)phenyl)ethanone
PC303395-10G PC303395-1G PC303395-5G PC303395-25G
APOSPC303395-10GEA 360 EUR
APOSPC303395-10G APOSPC303395-1G APOSPC303395-5G APOSPC303395-25G
4'-Chloro-2'-(trifluoromethyl)acetophenone 97%
4'-Chloro-2'-(trifluoromethyl)acetophenone
Formula: C₉H₆ClF₃O
MW: 222.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194258
CAS Number: 86520-34-5