6-Bromo-2,3-difluorophenylacetonitrile 97%
Supplier: Apollo Scientific
Synonyms: 2-(6-Bromo-2,3-difluorophenyl)acetonitrile
PC303433-1G PC303433-5G
APOSPC303433-1GEA 145 EUR
APOSPC303433-1G APOSPC303433-5G
6-Bromo-2,3-difluorophenylacetonitrile 97%
6-Bromo-2,3-difluorophenylacetonitrile
Formula: C₈H₄BrF₂N
MW: 232.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26686146
CAS Number: 1517576-71-4
UN: 3439
ADR: 6.1 III