2-Bromo-4,6-bis(trifluoromethyl)phenylacetonitrile
Supplier: Apollo Scientific
PC303531-1G PC303531-5G
APOSPC303531-1GEA 395 EUR
APOSPC303531-1G APOSPC303531-5G
2-Bromo-4,6-bis(trifluoromethyl)phenylacetonitrile
2-Bromo-4,6-bis(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₄BrF₆N
MW: 332.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28728179
CAS Number: 1805277-63-7