5-Bromo-2-fluoro-3-(trifluoromethyl)benzoyl chloride
Supplier: Apollo Scientific
PC303547-1G PC303547-5G
APOSPC303547-1GEA 244 EUR
APOSPC303547-1G APOSPC303547-5G
5-Bromo-2-fluoro-3-(trifluoromethyl)benzoyl chloride
5-Bromo-2-fluoro-3-(trifluoromethyl)benzoyl chloride
Formula: C₈H₂BrClF₄O
MW: 305.451 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640127
CAS Number: 2168482-19-5